4-Chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

20180-25G 20180-5G
76927-034EA 449.83 USD
76927-034 76927-036
4-Chloro-1H-indazole ≥97%
4-Chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00211065
CAS Number: 13096-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR