4-Fluoro-3-(tetrahydropyran-2-yloxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-(tetrahydro-2H-pyran-2-yloxy)phenylboronic acid

20144-1G 20144-25G 20144-5G
76927-026EA 73.21 USD
76927-026 76927-028 76927-030
4-Fluoro-3-(tetrahydropyran-2-yloxy)phenylboronic acid ≥97% research grade
4-Fluoro-3-(tetrahydropyran-2-yloxy)phenylboronic acid
Formula: C₁₁H₁₄BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD13195650
CAS Number: 1217501-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR