4-Bromo-6-chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

20104-25G 20104-5G
76927-000EA 244.01 USD
76927-000 76927-002
4-Bromo-6-chloro-1H-indazole ≥97%
4-Bromo-6-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781352
CAS Number: 885519-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR