4-Bromo-3-nitro-tert-butylbenzene ≥97%

Supplier: AOBChem USA

Warning

19950-1G 19950-25G 19950-500MG 19950-5G
76926-930EA 147.47 USD
76926-930 76926-932 76926-934 76926-936
4-Bromo-3-nitro-tert-butylbenzene ≥97%
4-Bromo-3-nitro-tert-butylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
Storage Temperature: Refrigerator
CAS Number: 70729-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR