4-Bromo-7-chloro-1H-indazole ≥97% research grade

Supplier: AOBChem USA

Warning

19884-1G 19884-25G 19884-5G
76926-910EA 94.43 USD
76926-910 76926-912 76926-914
4-Bromo-7-chloro-1H-indazole ≥97% research grade
4-Bromo-7-chloro-1H-indazole
Formula: C₇H₄BrClN₂
MW: 231.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16658654
CAS Number: 1186334-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR