5-Bromo-2-methylthioanisole ≥97%

Supplier: AOBChem USA

Warning

19850-1G 19850-25G 19850-500MG 19850-5G
76926-876EA 129.43 USD
76926-876 76926-878 76926-880 76926-882
5-Bromo-2-methylthioanisole ≥97%
5-Bromo-2-methylthioanisole

Precaución: for Research Use Only

Formula: C₈H₉BrS
Storage Temperature: Refrigerator
MDL Number: MFCD11855946
CAS Number: 142994-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR