4-Bromo-3-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-3-methoxyphenyl)methanol

19810-1G 19810-25G 19810-5G
76926-868EA 99.73 USD
76926-868 76926-870 76926-872
4-Bromo-3-methoxybenzyl alcohol ≥97%
4-Bromo-3-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11053671
CAS Number: 17100-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR