4-Bromo-3-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-3-methylphenyl)boronic acid

19735-1G 19735-25G 19735-5G
76926-848EA 62.6 USD
76926-848 76926-850 76926-852
4-Bromo-3-methylphenylboronic acid ≥97%
4-Bromo-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
MW: 214.852 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17011983
CAS Number: 221006-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR