4-Bromo-2-iodobenzonitrile ≥97% research grade

Supplier: AOBChem USA

Warning

19700-1G 19700-25G 19700-5G
76926-834EA 73.21 USD
76926-834 76926-836 76926-838
4-Bromo-2-iodobenzonitrile ≥97% research grade
4-Bromo-2-iodobenzonitrile
Formula: C₇H₃BrIN
Storage Temperature: Refrigerator
MDL Number: MFCD09907887
CAS Number: 944276-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR