1-Fluoro-2-iodo-4-methoxybenzene ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4-Fluoro-3-iodoanisole
19663-1G
19663-25G
19663-5G
76926-820EA
112.46
USD
76926-820
76926-822
76926-824
1-Fluoro-2-iodo-4-methoxybenzene ≥97% research grade
1-Fluoro-2-iodo-4-methoxybenzene
Formula:
C₇H₆FIO Storage Temperature: Refrigerator |
MDL Number:
MFCD16295276 CAS Number: 1028263-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...