4-Bromo-2,6-dichloroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

19515-25G 19515-5G
76926-786EA 209 USD
76926-786 76926-788
4-Bromo-2,6-dichloroiodobenzene ≥97%
4-Bromo-2,6-dichloroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂I
Storage Temperature: Ambient
CAS Number: 62720-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR