4-Hydroxymethyl-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Hydroxymethyl)-2-methoxyphenyl)boronic acid

19350-1G 19350-500MG 19350-5G
76926-750EA 468.92 USD
76926-750 76926-752 76926-754
4-Hydroxymethyl-2-methoxyphenylboronic acid ≥97%
4-Hydroxymethyl-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄
MW: 181.982 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295264
CAS Number: 1448869-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR