5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

19281-1G 19281-25G 19281-5G
76926-728EA 112.46 USD
76926-728 76926-730 76926-732
5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole ≥97%
5-Bromo-1-(2-tetrahydropyranyl)-1H-indazole
Formula: C₁₂H₁₃BrN₂O
MW: 281.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12922964
CAS Number: 478828-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR