4-Chloro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

19241-25G 19241-5G
76926-718EA 488.02 USD
76926-718 76926-720
4-Chloro-3-methylbenzaldehyde ≥97%
4-Chloro-3-methylbenzaldehyde
Formula: C₈H₇ClO
Storage Temperature: Ambient
MDL Number: MFCD02261736
CAS Number: 101349-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR