4-Chloro-3-(diethylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-(N,N-diethylcarbamoyl)phenylboronic acid

18957-1G 18957-25G 18957-500MG 18957-5G
76926-636EA 118.83 USD
76926-636 76926-638 76926-640 76926-642
4-Chloro-3-(diethylcarbamoyl)phenylboronic acid ≥97%
4-Chloro-3-(diethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD07363797
CAS Number: 871332-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR