5-Bromo-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

18945-25G 18945-5G
76926-624EA 244.01 USD
76926-624 76926-626
5-Bromo-1-methyl-1H-indazole ≥97%
5-Bromo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878568
CAS Number: 465529-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR