4-Chloro-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

18893-25G 18893-5G
76926-618EA 209 USD
76926-618 76926-620
4-Chloro-3-iodopyridine ≥97%
4-Chloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃ClIN
MW: 239.4415 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234024
CAS Number: 89167-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR