4-Methoxy-3-(methoxycarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

18633-1G 18633-25G 18633-500MG 18633-5G
76926-528EA 215.37 USD
76926-528 76926-530 76926-532 76926-534
4-Methoxy-3-(methoxycarbonyl)phenylboronic acid ≥97% research grade
4-Methoxy-3-(methoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₅
Storage Temperature: Ambient
MDL Number: MFCD23380361
CAS Number: 1071958-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR