4-Chloro-2,6-dimethybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18588-1G 18588-25G 18588-500MG 18588-5G
76926-510EA 165.51 USD
76926-510 76926-512 76926-514 76926-516
4-Chloro-2,6-dimethybenzaldehyde ≥97%
4-Chloro-2,6-dimethybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO
Storage Temperature: Refrigerator
MDL Number: MFCD11846957
CAS Number: 6045-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR