5-Bromo-2-chloro-N,N-dimethyl-3-pyridinecarboxamide ≥97%

Supplier: AOBChem USA

Warning

18553-1G 18553-25G 18553-500MG 18553-5G
76926-494EA 126.25 USD
76926-494 76926-496 76926-498 76926-500
5-Bromo-2-chloro-N,N-dimethyl-3-pyridinecarboxamide ≥97%
5-Bromo-2-chloro-N,N-dimethyl-3-pyridinecarboxamide

Precaución: for Research Use Only

Formula: C₈H₈BrClN₂O
MW: 263.519 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09746442
CAS Number: 1188142-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR