5-Chloro-3-fluoro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18377-1G 18377-25G 18377-500MG 18377-5G
76926-462EA 203.7 USD
76926-462 76926-464 76926-466 76926-468
5-Chloro-3-fluoro-2-hydroxybenzaldehyde ≥97%
5-Chloro-3-fluoro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.557 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16658025
CAS Number: 394-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR