5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
18308-1G
18308-25G
18308-5G
76926-430EA
84.88
USD
76926-430
76926-432
76926-434
5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine ≥97%
5-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine
Formula:
C₆H₃BrIN₃ Storage Temperature: Ambient |
MDL Number:
MFCD09878697 CAS Number: 875781-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...