5-Carboxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

18164-1G 18164-25G 18164-500MG 18164-5G
76926-394EA 153.84 USD
76926-394 76926-396 76926-398 76926-400
5-Carboxy-2-methylphenylboronic acid ≥97%
5-Carboxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
Storage Temperature: Ambient
MDL Number: MFCD08061639
CAS Number: 170230-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR