1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Acetylthiophene-2-boronic acid pinacol ester

18136-25G 18136-5G
76926-382EA 418 USD
76926-382 76926-384
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)ethanone ≥97%
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl)ethanone
Formula: C₁₂H₁₇BO₃S
MW: 252.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08063138
CAS Number: 942070-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR