4-Bromo-3,5-dimethylacetanilide ≥97%

Supplier: AOBChem USA

Warning

18125-25G 18125-5G
76926-370EA 346.92 USD
76926-370 76926-372
4-Bromo-3,5-dimethylacetanilide ≥97%
4-Bromo-3,5-dimethylacetanilide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD03931824
CAS Number: 64835-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR