4-Bromo-3,5-dimethylbenzonitrile ≥97% research grade

Supplier: AOBChem USA

Warning

18009-25G 18009-5G
76926-348EA 198.39 USD
76926-348 76926-350
4-Bromo-3,5-dimethylbenzonitrile ≥97% research grade
4-Bromo-3,5-dimethylbenzonitrile
Formula: C₉H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD07369910
CAS Number: 75344-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR