N-(4-Isopropyl-2-nitrophenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

17970-1G 17970-25G 17970-500MG 17970-5G
76926-334EA 146.41 USD
76926-334 76926-336 76926-338 76926-340
N-(4-Isopropyl-2-nitrophenyl)acetamide ≥97%
N-(4-Isopropyl-2-nitrophenyl)acetamide
Formula: C₁₁H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00094317
CAS Number: 40655-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR