5-Bromo-2-(trifluoromethoxy)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-trifluoromethoxyphenylboronic acid, (5-Bromo-2-(trifluoromethoxy)phenyl)boronic acid

17920-1G 17920-25G 17920-5G
76926-320EA 62.6 USD
76926-320 76926-322 76926-324
5-Bromo-2-(trifluoromethoxy)benzeneboronic acid ≥97%
5-Bromo-2-(trifluoromethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11504857
CAS Number: 1072951-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR