4-Carboxy-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-2-fluorophenylboronic Acid Pinacol Ester, 4-Carboxy-2-fluorobenzeneboronic acid pinacol ester

17682-25G 17682-5G
76926-264EA 381.93 USD
76926-264 76926-266
4-Carboxy-2-fluorophenylboronic acid pinacol ester ≥97%
4-Carboxy-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD12756466
CAS Number: 1050423-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR