5-Formyl-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17561-1G 17561-25G 17561-500MG 17561-5G
76926-214EA 350.1 USD
76926-214 76926-216 76926-218 76926-220
5-Formyl-3-methoxyphenylboronic acid ≥97%
5-Formyl-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD20526370
CAS Number: 1451392-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR