4-Cyano-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17444-1G 17444-25G 17444-500MG 17444-5G
76926-200EA 144.29 USD
76926-200 76926-202 76926-204 76926-206
4-Cyano-3-methoxyphenylboronic acid ≥97%
4-Cyano-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
Storage Temperature: Ambient
MDL Number: MFCD10696655
CAS Number: 677777-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR