5-Bromo-3-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

17417-1G 17417-25G 17417-5G
76926-180EA 70.02 USD
76926-180 76926-182 76926-184
5-Bromo-3-iodo-2-methoxypyridine ≥97%
5-Bromo-3-iodo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrINO
Storage Temperature: Refrigerator
MDL Number: MFCD05663522
CAS Number: 578007-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR