4-Bromo-2-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17381-1G 17381-25G 17381-5G
76926-162EA 84.88 USD
76926-162 76926-164 76926-166
4-Bromo-2-formylphenylboronic acid ≥97%
4-Bromo-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrO₃
MW: 228.836 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33403701
CAS Number: 2170041-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR