4-Fluoro-2-(methylthio)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

17179-1G 17179-25G 17179-500MG 17179-5G
76926-080EA 251.44 USD
76926-080 76926-082 76926-084 76926-086
4-Fluoro-2-(methylthio)phenylboronic acid ≥97% research grade
4-Fluoro-2-(methylthio)phenylboronic acid
Formula: C₇H₈BFO₂S
MW: 186.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18087696
CAS Number: 861931-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR