5-Bromo-2-(trifluoromethoxy)anisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-methoxy-1-(trifluoromethoxy)benzene

17099-1G 17099-25G 17099-500MG 17099-5G
76926-064EA 215.37 USD
76926-064 76926-066 76926-068 76926-070
5-Bromo-2-(trifluoromethoxy)anisole ≥97%
5-Bromo-2-(trifluoromethoxy)anisole
Formula: C₈H₆BrF₃O₂
MW: 271.031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21333068
CAS Number: 672948-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR