4-Bromo-3-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17066-1G 17066-25G 17066-500MG 17066-5G
76926-050EA 165.51 USD
76926-050 76926-052 76926-054 76926-056
4-Bromo-3-formylphenylboronic acid ≥97%
4-Bromo-3-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrO₃
MW: 228.836 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181642
CAS Number: 1451393-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR