5-(tert-Butyldimethylsilyloxy)-2-fluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (5-((tert-Butyldimethylsilyl)oxy)-2-fluorophenyl)boronic acid, 5-(t-Butyldimethylsilyloxy)-2-fluorophenylboronic acid

17047-1G 17047-25G 17047-500MG 17047-5G
76926-022EA 257.8 USD
76926-022 76926-024 76926-026 76926-028
5-(tert-Butyldimethylsilyloxy)-2-fluorophenylboronic acid ≥97% research grade
5-(tert-Butyldimethylsilyloxy)-2-fluorophenylboronic acid
Formula: C₁₂H₂₀BFO₃Si
MW: 270.1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855846
CAS Number: 1150114-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR