5-Bromo-4-methylnicotinic acid ≥95%

Supplier: AOBChem USA

Warning

17042-25G 17042-5G
76926-010EA 506.05 USD
76926-010 76926-012
5-Bromo-4-methylnicotinic acid ≥95%
5-Bromo-4-methylnicotinic acid
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07439973
CAS Number: 677702-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR