5-Bromo-3-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17004-1G 17004-25G 17004-500MG 17004-5G
76925-994EA 260.99 USD
76925-994 76925-996 76925-998 76926-000
5-Bromo-3-fluoro-2-methoxybenzaldehyde ≥97%
5-Bromo-3-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22035163
CAS Number: 470668-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR