4-Formyl-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16881-1G 16881-25G 16881-500MG 16881-5G
76925-954EA 177.18 USD
76925-954 76925-956 76925-958 76925-960
4-Formyl-3-methoxyphenylboronic acid ≥97%
4-Formyl-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
Storage Temperature: Ambient
MDL Number: MFCD10697427
CAS Number: 815620-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR