5-Ethoxy-2-fluorophenol ≥97% research grade

Supplier: AOBChem USA

Warning

16810-1G 16810-25G 16810-500MG 16810-5G
76925-918EA 217.49 USD
76925-918 76925-920 76925-922 76925-924
5-Ethoxy-2-fluorophenol ≥97% research grade
5-Ethoxy-2-fluorophenol
Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18974322
CAS Number: 577793-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR