4-Bromo-3,5-dimethylbenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

16694-1G 16694-25G 16694-5G
76925-880EA 62.6 USD
76925-880 76925-882 76925-884
4-Bromo-3,5-dimethylbenzaldehyde ≥97% research grade
4-Bromo-3,5-dimethylbenzaldehyde
Formula: C₉H₉BrO
MW: 213.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08063822
CAS Number: 400822-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR