5-Chloro-4-fluoro-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16589-1G 16589-500MG 16589-5G
76925-858EA 459.37 USD
76925-858 76925-860 76925-862
5-Chloro-4-fluoro-2-methoxyphenylboronic acid ≥97%
5-Chloro-4-fluoro-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
CAS Number: 949892-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR