5-Cyano-2-fluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Cyano-2-fluorophenylboronic acid pinacol ester, 4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

16562-1G 16562-25G 16562-5G
76925-834EA 79.57 USD
76925-834 76925-836 76925-838
5-Cyano-2-fluorobenzeneboronic acid pinacol ester ≥97%
5-Cyano-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₂
MW: 247.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07437899
CAS Number: 863868-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR