4-Bromo-2-ethoxy-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

16543-25G 16543-5G
76925-824EA 251.44 USD
76925-824 76925-826
4-Bromo-2-ethoxy-1-fluorobenzene ≥97%
4-Bromo-2-ethoxy-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17011974
CAS Number: 900174-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR