5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 3-borono-4-methoxybenzoate, 5-(Ethoxycarbonyl)-2-methoxyphenylboronic acid, 2-Borono-4-(ethoxycarbonyl)anisole

16519-1G 16519-25G 16519-5G
76925-790EA 100.79 USD
76925-790 76925-792 76925-794
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid ≥97%
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₅
Storage Temperature: Ambient
MDL Number: MFCD09800892
CAS Number: 957062-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR