5-Chloro-2-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Chloro-2-(trifluoromethyl)phenyl)methanol

16474-25G 16474-5G
76925-780EA 375.56 USD
76925-780 76925-782
5-Chloro-2-(trifluoromethyl)benzyl alcohol ≥97%
5-Chloro-2-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD01631355
CAS Number: 261763-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR