5-Chloro-2-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16472-25G 16472-5G
76925-772EA 219.61 USD
76925-772 76925-774
5-Chloro-2-(trifluoromethyl)benzaldehyde ≥97%
5-Chloro-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD03412204
CAS Number: 90381-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR