5-Bromo-2-chloronicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

16432-1G 16432-25G 16432-5G
76925-748EA 84.88 USD
76925-748 76925-750 76925-752
5-Bromo-2-chloronicotinaldehyde ≥97%
5-Bromo-2-chloronicotinaldehyde
Formula: C₆H₃BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD08689713
CAS Number: 228251-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR