4-Formyl-2-methylbenzeneboronic acid, pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-Formyl-2-methylphenylboronic acid pinacol ester
16421-1G
16421-25G
16421-500MG
16421-5G
76925-732EA
231.28
USD
76925-732
76925-734
76925-736
76925-738
4-Formyl-2-methylbenzeneboronic acid, pinacol ester ≥97%
4-Formyl-2-methylbenzeneboronic acid, pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BO₃ MW: 246.11 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08669583 CAS Number: 1073354-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...